Organic reference standard, Hexacosanoic acid, analytical standard, Supelco®
Catalog # 06922-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:506-46-7
- Description:Hexacosanoic acid
- Formula:C₂₆H₅₂O₂
- Storage temperature:2...8 °C
- Description:Hexacosanoic acid
Specifications
About this item
Hexacosanoic acid is classified under the very long chain fatty acids (VLCFAs) group of biologically important compounds.
- Identity confirmed by LC-MS method
Used as a reference standard for the determination of hexacosanoic acid in human serum and plasma samples for the screening of peroxisomal disorders by gas chromatography-mass spectrometry (GC-MS) operating under selected-ion monitoring (SIM) mode, in biological samples by GC-MS, in microalgal lipids by solid-phase extraction (SPE) followed by GC-MS analysis as a naphthoxyethyl derivative by high performance liquid chromatography (HPLC) coupled with fluorometric detection.