Order Entry
Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Furmecyclox
Supelco® PESTANAL® Organic Reference Standard, Furmecyclox
Catalog # 34347-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Furmecyclox
Catalog # 34347-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    60568-05-0
  • Description:
    Furmecyclox
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H351 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00466943
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N-Cyclohexyl-N-methoxy-2,5-dimethyl-3-furamide
  • Formula:
    C₁₄H₂₁NO₃
  • Molecular weight:
    251,32
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS08, GHS09
Caution: P201 - P273 - P308 + P313