Supelco® PESTANAL® Organic Reference Standard, Flurtamone
Catalog # 46286-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:96525-23-4
- Description:Flurtamone
- Storage conditions:2...30 °C
- Hazard class description:H302 - H311 - H410
- Shipping conditions:STD
- MDL No.:MFCD01725809
- M-Clarity quality level:MQ100
- Synonym(s):5-(Methylamino)-2-phenyl-4-[3-(trifluoromethyl)phenyl]-3(2H)-furanone
- Molecular weight:333,30
Specifications
About this item
- GHS signal: GHS09
Caution: P264 - P270 - P273 - P280 - P301 + P312 - P302 + P352 + P312