Supelco® PESTANAL® Organic Reference Standard, Flucythrinate
Catalog # 33496-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard, mixture of stereo isomers
- CAS No.:70124-77-5
- Description:Flucythrinate
- Storage conditions:2...8 °C
- Hazard class description:H226 - H301 - H332 - H410
- Shipping conditions:STD
- MDL No.:MFCD00137383
- M-Clarity quality level:MQ100
- Synonym(s):α-Cyano-3-phenoxybenzyl 4-difluoromethoxy-α-isopropylphenylacetate
- Formula:C₂₆H₂₃F₂NO₄
- Molecular weight:451,46
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P210 - P233 - P240 - P273 - P301 + P310 - P304 + P340 + P312