Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Fenofibric acid, analytical standard, Supelco®
Organic reference standard, Fenofibric acid, analytical standard, Supelco®
Catalog # 90568-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Fenofibric acid, analytical standard, Supelco®
Catalog # 90568-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    42017-89-0
  • Description:
    Fenofibric acid
  • Formula:
    C₁₇H₁₅ClO₄
  • Description:
    Fenofibric acid

Specifications

About this item

Used as a reference standard in determination of fenofibric acid in samples of human plasma using high performance liquid chromatography (HPLC) and liquid chromatography coupled with tandem mass spectrometry (LC-MS-MS).