Order Entry
Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, F.A.M.E. Mix, C4-C24
Supelco® Organic Reference Standard, F.A.M.E. Mix, C4-C24
Catalog # 18919-1AMP
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, F.A.M.E. Mix, C4-C24
Catalog # 18919-1AMP
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 Ampoul
  • Standards:
    Certified reference material, wt. % (varied)
  • Description:
    F.A.M.E. Mix, C4-C24
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H318 - H410
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    −20 °C
  • Pk Info:
    100 mg

Specifications

About this item

  • GHS signal: GHS05
Packaging: 100 mg ampule contains the following with weight percentages as indicated. Butyric acid methyl ester (C4:0) 4% Caproic acid methyl ester (C6:0) 4% Caprylic acid methyl ester (C8:0) 4% Capric acid methyl ester (C10:0) 4% Undecanoic acid methyl ester (C11:0) 2%
Lauric acid methyl ester (C12:0) 4%
Tridecanoic acid methyl ester (C13:0) 2%
Myristic acid methyl ester (C14:0) 4%
Myristoleic acid methyl ester (C14:1cis-9) 2%
Pentadecanoic acid methyl ester (C15:0) 2%
cis-10-Pentadecenoic acid methyl ester (15:1) 2%
Palmitic acid methyl ester (C16:0) 6%
Palmitoleic acid methyl ester (C16:1cis-9) 2%
Heptadecanoic acid methyl ester (C17:0) 2%
cis-10-Heptadecenoic acid methyl ester (C17:1) 2%
Stearic acid methyl ester (C18:0) 4%
Oleic acid methyl ester (C18:1cis-9) 4%
Elaidic acid methyl ester (C18:1 trans-9) 2%
Linoleic acid methyl ester (C18:2cis-912) 2%
Linolelaidic acid methyl ester (C18:2trans-912) 2%
Linolenic acid methyl ester (C18:3cis-91215) 2%
γ-Linolenic acid methyl ester (C18:3cis-6912) 2%
Arachidic acid methyl ester (C20:0) 4%
cis-11-Eicosenoic acid methyl ester (C20:1) 2%
cis-1114-Eicosadienoic acid methyl ester (C20:2) 2%
cis-111417-Eicosatrienoic acid methyl ester (C20:3) 2%
cis-81114-Eicosatrienoic acid methyl ester (C20:3) 2%
Arachidonic acid methyl ester (C20:4cis-581114) 2%
cis-58111417-Eicosapentaenoic acid methyl ester (C20:5) 2%
Heneicosanoic acid methyl ester (C21:0) 2%
Behenic acid methyl ester (C22:0) 4%
Erucic acid methyl ester (C22:1cis-13) 2%
cis-1316-Docosadienoic acid methyl ester (C22:2) 2%
cis-4710131619-Docosahexaenoic acid methyl ester (C22:6) 2
Caution: P273 - P280 - P305 + P351 + P338 - P391 - P501