Order Entry
Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Dichloroprop-P
Supelco® PESTANAL® Organic Reference Standard, Dichloroprop-P
Catalog # 31237-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Dichloroprop-P
Catalog # 31237-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    15165-67-0
  • Description:
    Dichloroprop-P
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H315 - H317 - H318
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00144036
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (R)-(+)-2-(2,4-Dichlorophenoxy)propionic acid
  • Formula:
    C₉H₈Cl₂O₃
  • Molecular weight:
    235,06
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS05, GHS07
Caution: P261 - P264 - P280 - P301 + P312 - P302 + P352 - P305 + P351 + P338