Supelco® PESTANAL® Organic Reference Standard, Coumatetralyl
Catalog # 45404-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:5836-29-3
- Description:Coumatetralyl
- Storage conditions:2...30 °C
- Hazard class description:H300 + H330 - H311 - H360D - H372 - H410
- Shipping conditions:STD
- MDL No.:MFCD09954502
- M-Clarity quality level:MQ100
- Formula:C₁₉H₁₆O₃
- Molecular weight:292,33
- Flash point:>100 °C
Specifications
About this item
- GHS signal: GHS08, GHS06
Caution: P202 - P260 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310