Order Entry
Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Chlorosulfuron
Supelco® PESTANAL® Organic Reference Standard, Chlorosulfuron
Catalog # 34322-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Chlorosulfuron
Catalog # 34322-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    64902-72-3
  • Description:
    Chlorosulfuron
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00128059
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-(2-Chlorophenylsulfonyl)-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-urea; Chlorsulfuron
  • Formula:
    C₁₂H₁₂ClN₅O₄S
  • Molecular weight:
    357,77
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS09
Caution: P273 - P391 - P501