Order Entry
Ireland
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Atrazine-D₅
 :  Merck
 
undefined
Supelco® PESTANAL® Organic Reference Standard, Atrazine-D₅
 :  Merck

 

  • CAS No.:
    19988-24-0
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₆H₁₁N₅O
  • Description:
    Atrazine-desethyl-2-hydroxy
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H317 - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00143637
  • Synonym(s):
    2-Chloro-4-pentadeuteroethylamino-6-isopropylamino-1,3,5-triazine
  • Molecular weight:
    220,71
  • Flash point:
    Not applicable

 

Frequently Bought Together