Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sinefungin
Sinefungin ≥95% (by HPLC), InSolution™, Calbiochem®, Sigma-Aldrich®
Sinefungin ≥95% (by HPLC), InSolution™, Calbiochem®, Sigma-Aldrich®
Sinefungin ≥95% (by HPLC), InSolution™, Calbiochem®, Sigma-Aldrich®
567051-2
:
Merck Millipore (Calbiochem)
:
Sinefungin ≥95% (by HPLC), InSolution™, Calbiochem®, Sigma-Aldrich®
567051-2
:
Merck Millipore (Calbiochem)
:
Pack type:
Plastic ampoule
Pk:
2 mg
Seq:
2145
Purity:
≥95% (HPLC)
:
:
Packaged under inert gas.
:
Freezer
:
58944-73-3