Test Strips, qualitative, silver
Catalog # MANA90732
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Pk:200
- Description:Silver ions Ag⁺
Specifications
About this item
The test paper allows the quick and easy detection of silver ions. These cause red-violet spots on a salmon background. The leaflet describes a method to eliminate interferences of Hg, Cu, Au, Pt and Pd.
- Colour changes: Salmon to red-violet
Limit of sensitivity: 20 mg/L Ag⁺
Packaging: Pack of 200 strips 20×70 mm.