Order Entry
Ireland
ContactUsLinkComponent
 
Pharmaceutical analytical standard, Diclazuril-methyl, reference material, Honeywell Fluka™
  HONCR1499-10MG
 :  
Pharmaceutical analytical standard, Diclazuril-methyl, reference material, Honeywell Fluka™
  HONCR1499-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    103337-71-9
  • Description:
    Diclazuril-methyl
  • Formula:
    C₁₈H₁₁Cl₃N₄O₂
  • Storage temperature:
    Ambient
  • Description:
    Diclazuril-methyl
  • Freezing point:
    181...182 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results