Organic analytical standards, Dicyandiamide-13C2,15N2, neat, reference material, Honeywell Fluka™
HONCR1490-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- Description:Dicyandiamide-¹³C₂,¹⁵N₂
- Formula:¹³C₂H₄N₂¹⁵N₂
- Storage temperature:Ambient
- Description:Dicyandiamide-¹³C₂,¹⁵N₂
- Freezing point:206...209 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results