Pesticide reference standard, Cycloprothrin, neat, Honeywell Fluka™
Catalog # HONCR1055-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:63935-38-6
- Description:Cycloprothrin
- Formula:C₂₆H₂₁Cl₂NO₄
- Storage temperature:2...8 °C
- Description:Cycloprothrin
- Freezing point:74...78 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results