Order Entry
Ireland
Orders LinkContactUsLinkComponent
Test Strips, Qualitative, Chromium

Test Strips, Qualitative, Chromium

Test paper chlortesmo for chlorine, 1 - mg/L - Cl₂, Color reaction with Michler's thioketone, 48×25×84 mm (packaging), 200 Tests, 200
Catalog # MANA90603
Supplier:   MACHEREY-NAGEL
Test Strips, Qualitative, Chromium
Catalog # MANA90603
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test Strips, Qualitative, Chromium
Test Strips, Qualitative, Chromium

Test paper chlortesmo for chlorine, 1 - mg/L - Cl₂, Color reaction with Michler's thioketone, 48×25×84 mm (packaging), 200 Tests, 200

CAS Number:  
Price
80,00 €
Log in for availability
1 * 200 items
Product Details & Documents

This test paper allows the quick and easy detection of chromate (CrO⁴₃). Cr(III) ions must be converted to Chromate prior to the detection.

  • Colour change: White to violet

For non-destructive testing of materials apply a drop of acid solution (1 part hydrochloric acid 37% + four parts hydrogen peroxide 3%) to the degreased surface. After 10 to 30 seconds add a drop sodium hydroxide (NaOH).

Specifications

Specifications

  • Chemical method:
    Color reaction with Michler's thioketone
  • Pk:
    200
  • Description:
    Test paper chlortesmo for chlorine
  • Dimensions:
    48×25×84 mm (packaging)
  • Tests per kit:
    200 Tests
  • Range:
    1 - mg/L - Cl₂
  • Cat. No.:
    MANA90603
  • Sensitivity:
    Chlorine - 1 mg/L Cl₂
  • Contents:
    200 test strips in a plastic box
  • Platform:
    Qualitative test paper
  • Compliance:
    Not CE certified and no hazardous material
  • Shelf life (months):
    24 (from production)
  • W×D×H:
    20×70 mm (net)
  • Method:
    Chlorine - pale-yellow → blue
  • Weight:
    50,64 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.