Malonyl coenzyme A lithium salt ≥90%
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:108347-84-8
:
Formula:C₂₄H₃₇N₇O₁₉P₃SLi
MDL Number:MFCD00057769
:859.6 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥90%
- λmax:257 nm
- Formulation:A Crystalline Solid
Specification Test Results
Frequently Bought Together