ESD safety ankle boots, lace-up, uvex 2 xenova®, 9556
Supplier: UVEX
Certificates
About this item
Metal-free, lightweight, flexible and robust safety shoe made from water-repellent, full-grain smooth leather upper material, soft padded tongue and collar, and a slip-resistant Xenova® dual-density PU sole resistant to heat, chemicals, oils and gasoline.
- Compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
- Anti-twist rear cap provides more stability and kink protection
- Climazone technology for improved comfort
- DUO-PUR sole, non-marking, with excellent cushioning and abrasion-resistant outsole
- Anti-static comfort insole with moisture control system; orthopaedic insoles
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Shock absorbing
- Fulfils the ESD requirement: Discharge resistance <35 megohm
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/blue
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S2, SRC
Frequently Bought Together