

Potassium chromate 1/30 M K2CrO4 (0.1N) volumetric solution, Fluka™
Catalog # 35157-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
About this item
:
:194.19 g/mol
Density:1 g/cm³
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:7789-00-6
Formula:K₂CrO₄
MDL Number:MFCD00011368