Test paper, chlorine
Catalog # MANA90709
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Pk:1
- Description:Chlorine test paper, 5 m roll
- Range:10 - 200 mg/l Cl₂
Specifications
About this item
The white paper turns dark blue in the presence of Cl₂. The test is particularly suitable for high range applications in disinfectant solutions which are for example used in food and beverage industry and animal farms.
- Long term stable and protected against external influences due to plastic packaging
- No accessories needed - always ready to use
- Quick and easy determination of chlorine
Colour reaction: White to dark blue