ViVi-CR Battery for Cleanroom Hood System
Supplier: THI Total Healthcare Innovation GmbH
Certificates
About this item
Black battery system with 49.87 Wh capacity. It charges faster, lasts longer, is equipped with a led display of charging state and ending battery acoustic alarm.
White small and lightweight Li-Ion battery pack with 43.6 Wh capacity. The charger holds four battery units.
Frequently Bought Together