Order Entry
Ireland
ContactUsLinkComponent
Ethidium homodimer I used in nuclear staining
Ethidium homodimer I used in nuclear staining
Catalog # 40010.
Supplier:  Biotium
CAS Number:  
undefined
Ethidium homodimer I used in nuclear staining
Catalog # 40010.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :856.75 g/mol
Storage Temperature:Refrigerator
UN:3096
ADR:8 (4.3),II
 :GHS08
 :GHS05
CAS Number:61926-22-5
Formula:C₄₆H₅₀Cl₄N₈
MDL Number:MFCD00210948