Dihydrorhodamine 123 ≥95% (by HPLC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:109244-58-8
:
Formula:C₂₁H₁₈N₂O₃
MDL Number:MFCD04221428
:346.386 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
Specification Test Results
- Appearance (Colour):Pink
- Appearance (Form):Powder
- Proton NMR Spectrum:Conforms to Structure
- Purity (HPLC):≥95%
Specification Test Results
Frequently Bought Together