Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Sand - pH 2.80, (CRM), Supelco®
Sand - pH 2.80, (CRM), Supelco®
Sand - pH 2.80, (CRM), Supelco®
Catalog # RTC000043-100G
Supplier:
Merck
CAS Number:
Expand image
Sand - pH 2.80, (CRM), Supelco®
Catalog # RTC000043-100G
Supplier:
Merck
CAS Number:
Specifications
Pk:
100 g
Description:
Sand - pH 2.80
Specifications
About this item
Product Details
Product Details