Supelco® TraceCERT® Organic Reference Standard, 2,2′-Methylenebis(6-tert-butyl-4-ethylphenol)
Catalog # 73096-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:88-24-4
- Description:2,2′-Methylenebis(6-tert-butyl-4-ethylphenol)
- Environmentally Preferable:
- Storage conditions:15...25 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00026296
- M-Clarity quality level:MQ300
- Melting point:119 - 122 °C (lit.)
- Formula:CH₂[C₆H₂[C(CH₃)₃](C₂H₅)OH]₂
- Molecular weight:368,55
Specifications
About this item
- GHS signal: GHS07
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338