0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, meropenem, (CRM), Supelco®
Pharmaceutical secondary standard, meropenem, (CRM), Supelco®
PHR1772-500MG
:
Merck
:
Pharmaceutical secondary standard, meropenem, (CRM), Supelco®
PHR1772-500MG
:
Merck
:
Pk:
500 mg
CAS No.:
119478-56-7
Description:
Meropenem
Description:
Meropenem