Supelco® TraceCERT® Organic Reference Standard, Dimethyl fumarate
50744-100MG
: Merck
:
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:624-49-7
- Description:Dimethyl fumarate
- Environmentally Preferable:
- Storage conditions:15...25 °C
- Hazard class description:H312 - H315 - H317 - H335 - H411
- Shipping conditions:STD
- MDL No.:MFCD00064438
- M-Clarity quality level:MQ300
- Melting point:102 - 106 °C (lit.)
- Formula:CH₃OCOCH=CHCOOCH₃
- Molecular weight:144,13
- Boiling point:192 - 193 °C (lit.)
- GHS signal: GHS07, GHS09
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P261 - P264 - P271 - P273 - P280 - P302 + P352 + P312