Drug and metabolite standards, HU-210 solution, (Spice Cannabinoid), (CRM), Supelco®
Catalog # S-024-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:112830-95-2
- Description:HU-210 solution, (Spice Cannabinoid)
- Description:HU-210 solution, (Spice Cannabinoid)
- Pk Info:ampoule of 1 ml