Order Entry
Ireland
ContactUsLinkComponent
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt (BCIP red)
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt (BCIP red)
Catalog # 10004.
Supplier:  Biotium
CAS Number:  
undefined
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt (BCIP red)
Catalog # 10004.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
CAS Number:6769-80-8
Formula:C₈H₆BrClNO₄P·C₇H₉N
MDL Number:MFCD00210023