Safety spectacles, uvex x-fit
Catalog # UVEX9199.085
Supplier: UVEX
Specifications
- Type:Anti-UV (UV 400), anti-scratch, anti-fog
- Colour:Grey
- Lenses:PC clear
- Pk:1
Specifications
About this item
The model x-fit are classic safety spectacles with good coverage of the eyes and a high level of wearer acceptance.
- Supravision coating technology
- Metal-free
- Translucent side arms
Weight: 23 g
Certifications: EN 166; EN 170