Order Entry
Ireland
ContactUsLinkComponent
Industrial gloves, uvex phynomic wet plus
 :  UVEX
undefined
Industrial gloves, uvex phynomic wet plus
 :  UVEX
  • Catalog No:
  • UVEX6006109
  • UVEX6006111
  • UVEX6006110
  • UVEX6006107
  • UVEX6006108
  • Model:
  • wet plus
  • wet plus
  • wet plus
  • wet plus
  • wet plus
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide/elastane
  • Polyamide/elastane
  • Polyamide/elastane
  • Polyamide/elastane
  • Polyamide/elastane
  • Finish:
  • Aqua-polymer foam coating on palm, fingertips and parts of the back of the hand
  • Aqua-polymer foam coating on palm, fingertips and parts of the back of the hand
  • Aqua-polymer foam coating on palm, fingertips and parts of the back of the hand
  • Aqua-polymer foam coating on palm, fingertips and parts of the back of the hand
  • Aqua-polymer foam coating on palm, fingertips and parts of the back of the hand
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
Frequently Bought Together