Chemical resistant gloves, uvex u-fit strong N2000
Supplier: UVEX
Certificates
About this item
These reinforced disposable gloves are made from 0,20 mm nitrile rubber and provide impressive mechanical strength. They are flexible and robust and provide good protection against many chemicals and splashes.
- Very high mechanical strength
- Outstanding tactile feel
- Good grip
- Silicone-free according to imprint test
Typical application include: Laboratories, chemical industry, precision assembly work, painting work, cleaning and food industry.
Certifications: EN ISO 374-1:2016 Type A (JKLOPST)
Food contact approved
Food contact approved
Specifications
- Catalog No:
- UVEX6096209
- UVEX6096210
- UVEX6096207
- UVEX6096211
- UVEX6096208
- Model:
- strong N2000
- strong N2000
- strong N2000
- strong N2000
- strong N2000
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Finish:
- Roughened fingertips
- Roughened fingertips
- Roughened fingertips
- Roughened fingertips
- Roughened fingertips
- EN norm:
- EN ISO 374-1:2016 Type A (JKLOPST)
- EN ISO 374-1:2016 Type A (JKLOPST)
- EN ISO 374-1:2016 Type A (JKLOPST)
- EN ISO 374-1:2016 Type A (JKLOPST)
- EN ISO 374-1:2016 Type A (JKLOPST)