Vial racks
Supplier: MACHEREY-NAGEL
Certificates
About this item
These chromatography vial racks are used for a safe standing position of the vials on the lab bench.
- For 1,5 and 4 ml vials or for 10 and 20 ml headspace vials
- Made from blue, chemically resistant PP
- Empty vial racks are stackable
Frequently Bought Together