Polystyrene-block-polybutadiene-block-polystyrene 30% (as styren), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9003-55-8
:
Formula:[CH₂CH(C₆H₅)]x(CH₂CH=CHCH₂)y [CH₂CH(C₆H₅)]x
MDL Number:MFCD00134010
Density:0,94 g/mL (25 °C) (lit.)
:M-Clarity™ quality level = MQ100
Specification Test Results
- Appearance (Color):Colorless to Yellow
- Appearance (Form):Beads
- Infrared spectrum:Conforms to Structure
- Viscosity 25 Wt% in Toluene at 25 °C:0.47 - 0.79 Pas
Specification Test Results
Frequently Bought Together