Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, fipronil, EP
Organic reference standard, fipronil, EP
Catalog # EDQMY0001968
Supplier:  EDQM
CAS Number:  
Organic reference standard, fipronil, EP
Catalog # EDQMY0001968
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    170 mg
  • Standards:
    Reference standard
  • CAS No.:
    120068-37-3
  • Description:
    Fipronil
  • Formula:
    C₁₂H₄Cl₂F₆N₄OS
  • Storage temperature:
    2…8 °C
  • Description:
    Fipronil

Specifications

About this item