Nitrile gloves, Showa 7555
Supplier: Showa
Certificates
About this item
These powder-free disposable gloves are made from 100% nitrile. They provide protection against the penetration and projection of chemicals.
- Low-modulus formulation to improve fit and reduce fatigue
- Chlorinated to offer more comfort, grip and reduce tackiness
- Textured finish on fingertips to enhance grip
- Second skin feel, softer texture and easy donning
- Latex-free to avoid type I and type IV latex allergies
Typical applications include: Aerospace, electronics, food, pharmaceutical and printing.
Certifications: EN 455, EN ISO 374-1 Type C (JK), EN ISO 374-5; Cat. III
Food contact approved.
Food contact approved.
Specifications
- Catalog No:
- BEST7555L
- BEST7555M
- BEST7555S
- BEST7555XL
- BEST7555XS
- BEST7555XXL
- Model:
- Showa 7555
- Showa 7555
- Showa 7555
- Showa 7555
- Showa 7555
- Showa 7555
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455, EN ISO 374-1 Type C (JK), EN ISO 374-5
- EN 455, EN ISO 374-1 Type C (JK), EN ISO 374-5
- EN 455, EN ISO 374-1 Type C (JK), EN ISO 374-5
- EN 455, EN ISO 374-1 Type C (JK), EN ISO 374-5
- EN 455, EN ISO 374-1 Type C (JK), EN ISO 374-5
- EN 455, EN ISO 374-1 Type C (JK), EN ISO 374-5
- Food contact approved:
- +
- +
- +
- +
- +
- +