Zinc oxide 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Catalog # 14439-1KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (calculated to ignited substance):99.0 - 100.5 %
- Loss on ignition (500°C):Max. 1.0 %
- Alkalinely reacting substances:complying
- Arsenic (As):Max. 3 ppm
- Cadmium (Cd):Max. 10 ppm
- Iron (Fe):Max. 10 ppm
- Lead (Pb):Max. 50 ppm
- Lead:complying
- Iron and other heavy metals:complying
- Carbonate and color of the solution:complying
- Carbonates and substances insoluble in acids:complying
- Residual solvents:complying
Specification Test Results
About this item
:
:81.39 g/mol
Melting Pt:1,975 °C
Density:5,6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-222-5
ADR:9,III
CAS Number:1314-13-2
Formula:ZnO
MDL Number:MFCD00011300