Order Entry
Ireland
ContactUsLinkComponent
Perchloric acid 70.0-72.0% (by alkalimetry), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Health
Corrosive
Irritant
Oxidizing
Perchloric acid 70.0-72.0% (by alkalimetry), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Catalog # 30755-2.5L
CAS Number:  
undefined
Health
Corrosive
Irritant
Oxidizing
Perchloric acid 70.0-72.0% (by alkalimetry), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Catalog # 30755-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (alkalimetric):
    70.0 - 72.0 %
  • Residue on ignition:
    Max. 0.003 %
  • Silver (Ag):
    Max. 0.00001 %
  • Aluminium (Al):
    Max. 0.000005 %
  • Arsenic (As):
    Max. 0.000005 %
  • Barium (Ba):
    Max. 0.000002 %
  • Beryllium (Be):
    Max. 0.000002 %
  • Bismuth (Bi):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000005 %
  • Copper (Cu):
    Max. 0.00001 %
  • Iron (Fe):
    Max. 0.0001 %
  • Potassium (K):
    Max. 0.00003 %
  • Lithium (Li):
    Max. 0.000002 %
  • Magnesium (Mg):
    Max. 0.00005 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Molybdenum (Mo):
    Max. 0.000005 %
  • Nickel (Ni):
    Max. 0.00001 %
  • Lead (Pb):
    Max. 0.000005 %
  • Strontium (Sr):
    Max. 0.000002 %
  • Titanium (Ti):
    Max. 0.00001 %
  • Thallium (Tl):
    Max. 0.000005 %
  • Vanadium (V):
    Max. 0.000005 %
  • Zinc (Zn):
    Max. 0.00001 %
  • Zirconium (Zr):
    Max. 0.00001 %
  • Heavy metals (as Pb):
    Max. 0.0001 %
  • Chlorate (ClO₃):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.0003 %
  • Free chlorine (Cl):
    Max. 0.00005 %
  • Total N:
    Max. 0.001 %
  • Phosphate, silicate (as SiO₂):
    Max. 0.0005 %
  • Sulfate (SO₄):
    Max. 0.001 %
  • APHA:
    Max. 10

Specification Test Results

About this item

 :
 :100.46 g/mol
Boiling Pt:203 °C
Melting Pt:-18 °C
Melting Pt:–18 °C
Density:1,66 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1873
ADR:5.1,I
 :GHS08
 :GHS05
 :GHS07
 :GHS03
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325