Order Entry
Ireland
ContactUsLinkComponent
Thymol blue indicator S. No.: 879, Fluka™
Thymol blue indicator S. No.: 879, Fluka™
Certificates
Thymol blue indicator S. No.: 879, Fluka™
Certificates
Specification Test Results
  • Identity (IR):
    complying
  • Change range (Ph1,2 - 2,8):
    complying
  • Change range (PH₈,0 - 9,6):
    complying
  • Indicator solution:
    complying
  • Extinction maximum at:
    589 - 599 nm
  • Extinction maximum of:
    700 - 900