Mercury(II) bromide ≥99.0% (by precipitation titration), Puriss. p.a. ACS, Fluka™
Catalog # 83353-50G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- Residue on ignition a. reducing with HCOOH (SO₄)*:Max. 0.02 %
- Insoluble in methanol:Max. 0.05 %
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 50 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 2000 ppm
- Sulfate (SO₄):Max. 50 ppm
- Ignition Residue (after reduction):≤0.02%
Specification Test Results
About this item
Synonyms: Mercuric bromide, Mercury dibromide
:
:360.4 g/mol
Merck Index:13,05900
Boiling Pt:322 °C
Melting Pt:237 °C
Density:6,1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-169-3
UN:1634
ADR:6.1,II
:GHS06
:GHS08
CAS Number:7789-47-1
Formula:Br₂Hg
MDL Number:MFCD00011040