Potassium thiocyanate ≥99.0% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Potassium rhodanide
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Merck Index:12,07866
Boiling Pt: Dec. (on heating)
Melting Pt:172 °C
Density:1,89 g/cm³
:97.18 g/mol
EINECS:206-370-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98.5 %
- Water insoluble matter:Max. 0.005 %
- pH (5 %, 25 °C):5.3 - 8.7
- Ammonium (NH₄):Max. 3 ppm
- Iron (Fe):Max. 2 ppm
- Sodium (Na):Max. 0.005 %
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 0.005 %
- Sulfate (SO₄):Max. 0.005 %
- Matter consuming I (as I):complying
- IR spectrum:complying
Specification Test Results
Frequently Bought Together