Order Entry
Ireland
ContactUsLinkComponent
Carbon disulphide ≥99.9%, Reagent Grade low in benzene
Carbon disulphide ≥99.9%, Reagent Grade low in benzene
Certificates
undefined
Carbon disulphide ≥99.9%, Reagent Grade low in benzene
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Explosion Limit:
    50 %
  • Assay (GC):
    Min. 99.90 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Non-volatile matter:
    Max. 0.0003 %
  • Benzene (GC):
    Max. 1 ppm
  • Autoignition Temperature:
    212 °F
  • Vapor Density:
    2.67 (vs air)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together