Potassium cyanide ≥97.0%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:151-50-8
:
Formula:KCN
MDL Number:MFCD00011397
:65.12 g/mol
Merck Index:13,07709
Melting Pt:634,5 °C
Density:1,56 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-792-3
UN:1680
ADR:6.1,I
:GHS06
Specification Test Results
- Assay:Min. 97 %
- Iron (Fe):Max. 0.01 %
- Sodium (Na):Max. 0.5 %
- Lead (Pb):Max. 0.0002 %
- Chloride (Cl):Max. 0.02 %
- Phosphate (PO₄):Max. 0.005 %
- Sulfate (SO₄):Max. 0.04 %
- Sulfide (S):Max. 0.0005 %
- Thiocyanate (SCN):Max. 0.01 %
Specification Test Results
Frequently Bought Together