
Ethylene glycol ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:107-21-1
:
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885
:62.07 g/mol
Merck Index:13,03832
Boiling Pt:197 °C
Melting Pt:–12 °C
Density:1,11 g/cm³ (20 °C)
Flash Pt:111 °C
Storage Temperature:Ambient
EINECS:203-473-3
REACH:01-2119456816-28
:GHS08
Specification Test Results
- Refractive Index:n20/D 1.431 (lit.)
- Assay (GC):Min. 99.0 %
- IR spectrum:complying
- APHA:Max. 10
- Vapor Density:2.1 (vs air)
- Explosion Limit:15.3 %
Specification Test Results
Frequently Bought Together