


1-Propanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
:71-23-8
:
:H₃CCH₂CH₂OH
:MFCD00002941
:60.1 g/mol
:13,07934
:96…98 °C (1,013 hPa)
:–127 °C
:0,803…0.804 g/cm³ (20 °C)
:10,8 %(V)
:Ambient
:200-746-9
:1274
:3,II
:GHS07
:GHS05
:GHS02
- Assay (GC):Min. 99.5 %
- Boiling range:96 - 99 °C
- Density (D 20/20):0.802 - 0.806
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.05 %
- Free acid (as C₂H₅COOH):Max. 0.001 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- Acetone (GC):Max. 0.01 %
- Ethanol (GC):Max. 0.05 %
- Methanol (GC):Max. 0.05 %
- 2-propanol (GC):Max. 0.05 %
- Propionaldehyde (GC):Max. 0.01 %
- Autoignition Temperature:700 °F
Frequently Bought Together