Order Entry
Ireland
ContactUsLinkComponent
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Health
Toxic
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Catalog # 319988-500ML
CAS Number:  
undefined
Health
Toxic
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Catalog # 319988-500ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Vapor Pressure:
    160 mmHg ( 20 °C)
  • Identity (IR):
    complying
  • Purity excluding stabilizer (GC):
    Min. 99.8 %
  • Ethanol (GC):
    0.5 - 1.0 Gew.%
  • Non-volatile matter:
    Max. 0.001 %
  • Lead (Pb):
    Max. 0.05 ppm
  • Free chlorine:
    complying
  • Free acid and chloride:
    complying
  • Acetone, aldehydes (neßler test):
    complying
  • Reaction against H₂SO₄:
    complying
  • APHA:
    Max. 10
  • Vapor Density:
    4.1 (vs air)

Specification Test Results

About this item

 :
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:60-62 °C (1,013 hPa)
Boiling Pt:60…62 °C (1,013 hPa)
Melting Pt:-63 °C
Melting Pt:–63 °C
Density:1,48 g/cm3 (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
 :Stabilised with ethanol 0,5-1,0%
REACH:05-2114614906-45
 :GHS08
 :GHS06
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826