Order Entry
Ireland
ContactUsLinkComponent
Analytical standard, Posaconazol, reference material, Honeywell Fluka™
Analytical standard, Posaconazol, reference material, Honeywell Fluka™
  HONCR1316-25MG
 :  
Analytical standard, Posaconazol, reference material, Honeywell Fluka™
  HONCR1316-25MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    25 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    171228-49-2
  • Description:
    Posaconazol
  • Melting point:
    168…174 °C
  • Formula:
    C₃₇H₄₂F₂N₈O₄
  • Storage temperature:
    Ambient
  • Description:
    Posaconazol

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results