Order Entry
Ireland
ContactUsLinkComponent
Analytical standard, Trifluralin-(dipropyl-d14), reference material, Honeywell Fluka™
Analytical standard, Trifluralin-(dipropyl-d14), reference material, Honeywell Fluka™
Catalog # HONCR1304-10MG
CAS Number:  
Analytical standard, Trifluralin-(dipropyl-d14), reference material, Honeywell Fluka™
Catalog # HONCR1304-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    347841-79-6
  • Description:
    Trifluralin-(dipropyl-d14)
  • Melting point:
    40…45 °C
  • Formula:
    C₁₃D₁₄H₂F₃N₃O₄
  • Storage temperature:
    Ambient
  • Description:
    Trifluralin-(dipropyl-d14)

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results