Analytical standard, (EZ)-Cyenopyrafen, reference material, Honeywell Fluka™
Catalog # HONCR1203-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:268743-66-4
- Description:(EZ)-Cyenopyrafen
- Formula:C₂₄H₃₁N₃O₂
- Storage temperature:Ambient
- Description:(EZ)-Cyenopyrafen
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results