Order Entry
Ireland
ContactUsLinkComponent
Analytical standard, Chlorimuron ethyl, reference material, Honeywell Fluka™
Analytical standard, Chlorimuron ethyl, reference material, Honeywell Fluka™
  HONCR1138-100MG
 :  
Analytical standard, Chlorimuron ethyl, reference material, Honeywell Fluka™
  HONCR1138-100MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    90982-32-4
  • Description:
    Chlorimuron ethyl
  • Melting point:
    181 °C
  • Formula:
    C₁₅H₁₅ClN₄O₆S
  • Storage temperature:
    Ambient
  • Description:
    Chlorimuron ethyl

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results